Anti-CX3CR1 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: SA011F11]

Supplier: Biolegend
149003 149004
CA10756-800EA 253.73 CAD
CA10756-800 CA10756-802
Anti-CX3CR1 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: SA011F11]
Antibodies
Alexa Fluor® 647 anti-mouse CX3CR1 [SA011F11]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CX3CR1
Clonality: Monoclonal
Clone: SA011F11
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR