Anti-CD95 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: DX2]

Supplier: Biolegend
305633 305634
CA10757-936EA 234.58 CAD
CA10757-936 CA10757-938
Anti-CD95 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: DX2]
Antibodies
PE/Dazzle™ 594 anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD95
Clonality: Monoclonal
Clone: DX2
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Chimpanzee;Common Marmoset;Cotton-topped Tamarin;Cynomolgus;Pigtailed Macaque;Rhesus;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR