Anti-CD90.2 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 53-2.1]

Supplier: Biolegend
140305 140306
CA140305-BLEA 161.18 CAD
CA140305-BL CA140306-BL
Anti-CD90.2 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 53-2.1]
Antibodies
Pacific Blue™ anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 53-2.1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR