Anti-CD90.2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 53-2.1]

Supplier: Biolegend
140303 140304
CA140303-BLEA 86.17 CAD
CA140303-BL CA140304-BL
Anti-CD90.2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 53-2.1]
Antibodies
FITC anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 53-2.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR