Anti-CD8a Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 53-6.7]

Supplier: Biolegend
100747 100748 100759
CA100747-BLEA 333.52 CAD
CA100747-BL CA10756-698 CA10756-710
Anti-CD8a Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 53-6.7]
Antibodies
Brilliant Violet 711™ anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD8a CD8a
Environmentally Preferable
Antigen synonyms T8|Lyt 2|Lyt2|Ly 2|Ly2|Ly-2!Lyt-2|CD8 T8|Lyt 2|Lyt2|Ly 2|Ly2|Ly-2!Lyt-2|CD8
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation Brilliant Violet® 711 Brilliant Violet® 711
Clone 53-6.7 53-6.7
Reactivity Mouse Mouse
Host Rat Rat
Gene ID 12525 12525
Isotype IgG2a kappa IgG2a kappa
Epitope α chain α chain
Immunogen Mouse thymus or spleen Mouse thymus or spleen
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA). Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 711™ and unconjugated antibody. The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 711™ and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze. The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR