Anti-CD80 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 16-10A1]

Supplier: Biolegend
104737 104738
CA10756-866EA 225.01 CAD
CA10756-866 CA10756-868
Anti-CD80 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 16-10A1]
Antibodies
PE/Dazzle™ 594 anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Cross-Reactivity: Dog (Canine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR