Anti-CD80 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 16-10A1]

Supplier: Biolegend
104733 104734
CA10756-860EA 185.11 CAD
CA10756-860 CA10756-862
Anti-CD80 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 16-10A1]
Antibodies
PE/Cy7 anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Cross-Reactivity: Dog (Canine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR