Anti-CD69 Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: H1.2F3]

Supplier: Biolegend
104529 104530
CA104529-BLEA 343.1 CAD
CA104529-BL CA10756-852
Anti-CD69 Armenian Hamster Monoclonal Antibody (Brilliant Violet® 605) [clone: H1.2F3]
Antibodies
Brilliant Violet 605™ anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Brilliant Violet® 605
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR