Anti-CD66b Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: G10F5]

Supplier: Biolegend
305111 305112
CA10757-892EA 217.03 CAD
CA10757-892 CA10757-894
Anti-CD66b Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: G10F5]
Antibodies
Pacific Blue™ anti-human CD66b [G10F5]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD66b
Clonality: Monoclonal
Clone: G10F5
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR