Anti-CD64 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: X54-5/7.1]

Supplier: Biolegend
139313 139314
CA10757-200EA 280.86 CAD
CA10757-200 CA10757-202
Anti-CD64 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: X54-5/7.1]
Antibodies
PE/Cy7 anti-mouse CD64 (FcγRI) [X54-5/7.1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: X54-5/7.1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR