Anti-CD57 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HNK-1]

Supplier: Biolegend
359607 359608
CA10760-630EA 197.88 CAD
CA10760-630 CA10760-632
Anti-CD57 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HNK-1]
Antibodies
Pacific Blue™ anti-human CD57 [HNK-1]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: HNK-1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR