Anti-CD49b Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1E6-C5]

Supplier: Biolegend
359307 359308
CA10760-392EA 194.69 CAD
CA10760-392 CA10760-394
Anti-CD49b Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1E6-C5]
Antibodies
PE anti-human CD49b [P1E6-C5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49b
Clonality: Monoclonal
Clone: P1E6-C5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR