Anti-CD49a Armenian Hamster Monoclonal Antibody [clone: HMalpha1]

Supplier: Biolegend
142601 142602
CA142601-BLEA 122.88 CAD
CA142601-BL CA142602-BL
Anti-CD49a Armenian Hamster Monoclonal Antibody [clone: HMalpha1]
Antibodies
Purified anti-mouse CD49a [HMα1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49a
Clonality: Monoclonal
Clone: HMalpha1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR