Anti-CD45RO Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: UCHL1]

Supplier: Biolegend
304233 304234
CA304233-BLEA 368.63 CAD
CA304233-BL CA10757-832
Anti-CD45RO Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: UCHL1]
Antibodies
Brilliant Violet 785™ anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCHL1
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Chimpanzee;Cynomolgus;Common Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR