Anti-CD45RA Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: HI100]

Supplier: Biolegend
304139 304140
CA10757-586EA 343.1 CAD
CA10757-586 CA10757-588
Anti-CD45RA Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: HI100]
Antibodies
Brilliant Violet 785™ anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: HI100
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR