Anti-CD45R Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: RA3-6B2]
Supplier: Biolegend
103243 103244
CA103243-BLEA 333.52 CAD
CA103243-BL CA10756-830
Anti-CD45R Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: RA3-6B2]
Antibodies
Brilliant Violet 605™ anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC; Size: 50 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent beMore Product Information