Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: A20]

Supplier: Biolegend
110747 110748
CA10756-360EA 225.01 CAD
CA10756-360 CA10756-362
Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: A20]
Antibodies
PE/Dazzle™ 594 anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse (A.SW) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR