Anti-CD45 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: I3/2.3]

Supplier: Biolegend
147707 147708
CA10756-614EA 124.47 CAD
CA10756-614 CA10756-616
Anti-CD45 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: I3/2.3]
Antibodies
APC anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: I3/2.3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR