Anti-CD45 Rat Monoclonal Antibody [clone: I3/2.3]

Supplier: Biolegend
147701 147702
CA10756-602EA 86.17 CAD
CA10756-602 CA10756-604
Anti-CD45 Rat Monoclonal Antibody [clone: I3/2.3]
Antibodies
Purified anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: I3/2.3
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR