Anti-CD45 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: HI30]

Supplier: Biolegend
304044 304043
CA10757-574EA 639.92 CAD
CA10757-574 CA10757-812
Anti-CD45 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: HI30]
Antibodies
Brilliant Violet 650™ anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: HI30
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR