Anti-CD43 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: CD43-10G7]

Supplier: Biolegend
343207 343208
CA10758-678EA 178.73 CAD
CA10758-678 CA10758-680
Anti-CD43 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: CD43-10G7]
Antibodies
PE/Cy7 anti-human CD43 [CD43-10G7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: CD43-10G7
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR