Anti-CD43 Rat Monoclonal Antibody [clone: S11]

Supplier: Biolegend
143201 143202
CA143201-BLEA 103.73 CAD
CA143201-BL CA143202-BL
Anti-CD43 Rat Monoclonal Antibody [clone: S11]
Antibodies
Purified anti-mouse CD43 [S11]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: S11
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR