Anti-CD41/CD61 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: PAC-1]

Supplier: Biolegend
362803 362804
CA10760-910EA 234.58 CAD
CA10760-910 CA10760-912
Anti-CD41/CD61 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: PAC-1]
Antibodies
FITC anti-human CD41/CD61 [PAC-1]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD41/CD61
Clonality: Monoclonal
Clone: PAC-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD41/CD61
Environmentally Preferable
Antigen synonyms glycoprotein IIb/IIIa complex|integrin αIIbβ3|gpIIb/IIIa complex
Antibody type Primary
Clonality Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Clone PAC-1
Reactivity Human
Host Mouse
Gene ID 3674, 3690
Isotype IgM kappa
Immunogen Human platelets
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR