Anti-CD41 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIP8]

Supplier: Biolegend
303713 303714
CA303713-BLEA 253.73 CAD
CA303713-BL CA303714-BL
Anti-CD41 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIP8]
Antibodies
Pacific Blue™ anti-human CD41 [HIP8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: HIP8
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR