Anti-CD40 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 5C3]

Supplier: Biolegend
334335 334336
CA10758-534EA 343.1 CAD
CA10758-534 CA10758-536
Anti-CD40 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 5C3]
Antibodies
Brilliant Violet 605™ anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 5C3
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Baboon;Chimpanzee;Cynomolgus Monkey;Rhesus;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR