Anti-CD40 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 5C3]

Supplier: Biolegend
334321 334322
CA334321-BLEA 225.01 CAD
CA334321-BL CA334322-BL
Anti-CD40 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 5C3]
Antibodies
PE/Cy7 anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for eachMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD40
Environmentally Preferable
Antigen synonyms BP50|TNFRSF5
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)/Cy7®
Clone 5C3
Reactivity Baboon,Cynomolgus monkey,Rhesus monkey,Chimpanzee,Human,Squirrel monkey
Host Mouse
Gene ID 958
Isotype IgG1 kappa
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR