Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: RPA-T4]

Supplier: Biolegend
300558 300557
CA10757-486EA 667.04 CAD
CA10757-486 CA10757-564
Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: RPA-T4]
Antibodies
Brilliant Violet 711™ anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR