Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RM4-4]

Supplier: Biolegend
116013 116014
CA10756-944EA 124.47 CAD
CA10756-944 CA10756-946
Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RM4-4]
Antibodies
APC anti-mouse CD4 [RM4-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR