Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: OKT4]

Supplier: Biolegend
317433 317434
CA317433-BLEA 263.31 CAD
CA317433-BL CA10758-200
Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: OKT4]
Antibodies
Brilliant Violet 421™ anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: Brilliant Violet® 421
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR