Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: HIT2]

Supplier: Biolegend
303533 303534
CA10757-782EA 244.16 CAD
CA10757-782 CA10757-784
Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: HIT2]
Antibodies
APC/Cy7 anti-human CD38 [HIT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Horse, Cow (Bovine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Horse;Cow (Bovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR