Anti-CD38 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: HIT2]

Supplier: Biolegend
303527 303528
CA303527-BLEA 368.63 CAD
CA303527-BL CA10757-778
Anti-CD38 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: HIT2]
Antibodies
Brilliant Violet 711™ anti-human CD38 [HIT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Horse, Cow (Bovine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Horse;Cow (Bovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR