Anti-CD371 (CLEC12A) Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 50C1]

Supplier: Biolegend
353607 353608
CA10759-410EA 188.3 CAD
CA10759-410 CA10759-412
Anti-CD371 (CLEC12A) Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 50C1]
Antibodies
FITC anti-human CD371 (CLEC12A) [50C1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD371 (CLEC12A)
Clonality: Monoclonal
Clone: 50C1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR