Anti-CD366 (Tim-3) Mouse Monoclonal Antibody [clone: F38-2E2]

Supplier: Biolegend
345009 345010
CA345009-BLEA 430.87 CAD
CA345009-BL CA345010-BL
Anti-CD366 (Tim-3) Mouse Monoclonal Antibody [clone: F38-2E2]
Antibodies
Ultra-LEAF™ Purified anti-human CD366 (Tim-3) [F38-2E2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC, Costim, Block; Size: 1 mg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD366 (Tim-3)
Clonality: Monoclonal
Clone: F38-2E2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR