Anti-CD360 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A12]

Supplier: Biolegend
359505 359506
CA10760-610EA 232.99 CAD
CA10760-610 CA10760-612
Anti-CD360 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A12]
Antibodies
PE anti-human CD360 (IL-21R) [17A12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD360
Clonality: Monoclonal
Clone: 17A12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR