Anti-CD326 (Ep-CAM) Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: 9C4]

Supplier: Biolegend
324225 324226
CA10758-294EA 343.1 CAD
CA10758-294 CA10758-296
Anti-CD326 (Ep-CAM) Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: 9C4]
Antibodies
Brilliant Violet 650™ anti-human CD326 (Ep-CAM) [9C4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD326 (Ep-CAM)
Clonality: Monoclonal
Clone: 9C4
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR