Anti-CD3 Rat Monoclonal Antibody (Brilliant Violet® 785) [clone: 17A2]

Supplier: Biolegend
100231 100232
CA100231-BLEA 351.08 CAD
CA100231-BL CA10756-654
Anti-CD3 Rat Monoclonal Antibody (Brilliant Violet® 785) [clone: 17A2]
Antibodies
Brilliant Violet 785™ anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: Brilliant Violet® 785
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR