Anti-CD3 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: OKT3]

Supplier: Biolegend
317323 317324
CA317323-BLEA 343.1 CAD
CA317323-BL CA10758-176
Anti-CD3 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: OKT3]
Antibodies
Brilliant Violet 650™ anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR