Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1]

Supplier: Biolegend
300437 300438
CA300437-BLEA 151.6 CAD
CA300437-BL CA300438-BL
Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1]
Antibodies
Ultra-LEAF™ Purified anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, CyTOF®, IP, Activ, IHC, WB; Size: 1 mg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR