Anti-CD285 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: ACT5]

Supplier: Biolegend
148103 148104
CA10756-632EA 253.73 CAD
CA10756-632 CA10756-634
Anti-CD285 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: ACT5]
Antibodies
Alexa Fluor® 647 anti-mouse CD285 (TLR5) [ACT5]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD285
Clonality: Monoclonal
Clone: ACT5
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR