Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2]

Supplier: Biolegend
302933 302934
CA302933-BLEA 175.54 CAD
CA302933-BL CA302934-BL
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2]
Antibodies
Ultra-LEAF™ Purified anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC, Activ, Costim, IHC, IP; Size: 1 mg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus;Sooty Mangabey;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR