Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: C398.4A]

Supplier: Biolegend
313523 313524
CA10758-120EA 315.97 CAD
CA10758-120 CA10758-122
Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: C398.4A]
Antibodies
Brilliant Violet 421™ anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD278 (ICOS)
Clonality: Monoclonal
Clone: C398.4A
Conjugation: Brilliant Violet® 421
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Human;African Green;Baboon;Cynomolgus;Mouse;Rat;Rhesus;Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR