Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: C398.4A]

Supplier: Biolegend
313519 313520
CA313519-BLEA 169.15 CAD
CA313519-BL CA313520-BL
Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: C398.4A]
Antibodies
PE/Cy7 anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD278 (ICOS)
Clonality: Monoclonal
Clone: C398.4A
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR