Anti-CD25 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: PC61]

Supplier: Biolegend
102041 102042
CA10756-766EA 307.99 CAD
CA10756-766 CA10756-768
Anti-CD25 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: PC61]
Antibodies
Brilliant Violet 510™ anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: PC61
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: Rat IgG1, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR