Anti-CD235a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HI264]

Supplier: Biolegend
349107 349108
CA349107-BLEA 234.58 CAD
CA349107-BL CA349108-BL
Anti-CD235a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HI264]
Antibodies
Pacific Blue™ anti-human CD235a (Glycophorin A) [HI264]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD235a
Clonality: Monoclonal
Clone: HI264
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR