Anti-CD22 Mouse Monoclonal Antibody [clone: S-HCL-1]

Supplier: Biolegend
363502 363501
CA10760-990EA 217.03 CAD
CA10760-990 CA10761-060
Anti-CD22 Mouse Monoclonal Antibody [clone: S-HCL-1]
Antibodies
Purified anti-human CD22 [S-HCL-1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: S-HCL-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR