Anti-CD213a2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 47]

Supplier: Biolegend
360305 360306
CA10760-726EA 244.16 CAD
CA10760-726 CA10760-728
Anti-CD213a2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 47]
Antibodies
PE anti-human CD213α2 (IL13Rα2) [47]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD213a2
Clonality: Monoclonal
Clone: 47
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD213a2
Environmentally Preferable
Antigen synonyms IL13RA2|CD213a2|IL-13R|CT19|IL13BP|IL-13RA2
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone 47
Reactivity Human
Host Mouse
Gene ID 3598
Isotype IgG1 kappa
Immunogen Recombinant human IL-13R alpha 2 Fc chimera
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR