Anti-CD200 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: OX-104]

Supplier: Biolegend
329217 329218
CA10758-386EA 325.54 CAD
CA10758-386 CA10758-388
Anti-CD200 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: OX-104]
Antibodies
Brilliant Violet 605™ anti-human CD200 (OX2) [OX-104]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: OX-104
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR