Anti-CD194 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: L291H4]

Supplier: Biolegend
359417 359418
CA10760-598EA 315.97 CAD
CA10760-598 CA10760-600
Anti-CD194 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: L291H4]
Antibodies
Brilliant Violet 605™ anti-human CD194 (CCR4) [L291H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD194
Clonality: Monoclonal
Clone: L291H4
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR