Anti-CD19 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: SJ25C1]

Supplier: Biolegend
363025 363026
CA10761-040EA 320.76 CAD
CA10761-040 CA10761-042
Anti-CD19 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: SJ25C1]
Antibodies
Brilliant Violet 650™ anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR