Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1]

Supplier: Biolegend
363001 363002
CA10760-928EA 86.17 CAD
CA10760-928 CA10760-930
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1]
Antibodies
Purified anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD19
Environmentally Preferable
Antigen synonyms B-lymphocyte Antigen CD19|T-cell Surface Antigen Leu-12|CVID3|SJ25-C1|CD19 Antigen|CD19 Molecule|B-Lymphocytes Surface Antigen B4|Differentiation Antigen CD19|B4
Antibody type Primary
Clonality Monoclonal
Clone SJ25C1
Reactivity Human
Host Mouse
Gene ID 930
Isotype IgG1 kappa
ImmunoChemistry Yes
Immunogen NALM1 + NALM16 cells
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR